tert-Butyl iodoacetate 97%

Supplier: Ambeed
Danger

A329520-100MG A329520-1G A329520-250MG A329520-5G A329520-10G A329520-25G
76803-022EA 11 USD
76803-022 76803-024 76803-026 77668-490 77879-697 77879-698
tert-Butyl iodoacetate 97%
tert-Butyl iodoacetate

Precaución: for Research Use Only

Formula: C₆H₁₁IO₂
MDL Number: MFCD09833728
CAS Number: 49827-15-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR