Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate 96%

Supplier: Ambeed
Warning

A328795-1G A328795-250MG
76803-010EA 54.11 USD
76803-010 76803-012
Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate 96%
Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.226 g/mol
MDL Number: MFCD03981003
CAS Number: 57060-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR