Fmoc-Asp(OtBu)-OL 95%

Supplier: Ambeed
Warning

Synonyms: AmbotzFAL3060, Fmoc-Aspartimol(OtBu)

A328403-1G A328403-250MG A328403-5G
76803-000EA 92.3 USD
76803-000 76803-002 76803-004
Fmoc-Asp(OtBu)-OL 95%
Fmoc-Asp(OtBu)-OL

Precaución: for Research Use Only

Formula: C₂₃H₂₇NO₅
MW: 397.46 g/mol
MDL Number: MFCD00065675
CAS Number: 133565-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR