Imidazo[1,5-a]pyridine-6-carboxylic acid 98%

Supplier: Ambeed
Warning

A326902-1G A326902-250MG A326902-100MG
76802-992EA 240 USD
76802-992 76802-994 77668-478
Imidazo[1,5-a]pyridine-6-carboxylic acid 98%
Imidazo[1,5-a]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD11042992
CAS Number: 256935-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR