2-(2-Propynyloxy)benzenecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Prop-2-yn-1-yloxy)benzaldehyde, 2-(2-Propynyloxy)benzaldehyde, 2-(Propargyloxy)benzaldehyde

A326749-250MG A326749-5G A326749-1G
76802-990EA 10.61 USD
76802-990 77550-906 77626-612
2-(2-Propynyloxy)benzenecarbaldehyde 98%
2-(2-Propynyloxy)benzenecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
MDL Number: MFCD02380378
CAS Number: 29978-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR