Ethyl-3-hexenoate 95%

Supplier: Ambeed
Danger

Synonyms: Ethyl hex-3-enoate

A326264-25G A326264-5G
76802-986EA 16.98 USD
76802-986 76802-988
Ethyl-3-hexenoate 95%
Ethyl-3-hexenoate

Precaución: for Research Use Only

Formula: CH₃CH₂CH=CHCH₂CO₂CH₂CH₃
MW: 142.20 g/mol
MDL Number: MFCD00036524
CAS Number: 2396-83-0
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR