6-Hydroxy-2-naphthaleneboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(6-Hydroxynaphthalen-2-yl)boronic acid, 6-Hydroxynaphthalene-2-boronic acid
A325171-100MG
A325171-1G
A325171-250MG
A325171-25G
A325171-5G
76802-960EA
5.31
USD
76802-960
76802-962
76802-964
76802-966
76802-968
6-Hydroxy-2-naphthaleneboronic acid 98%
6-Hydroxy-2-naphthaleneboronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉BO₃ MW: 187.99 g/mol |
MDL Number:
MFCD00092986 CAS Number: 173194-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...