3-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)quinazoline-2,4(1H,3H)-dione (2R,3R)-2,3-dihydroxysuccinate 97%
Supplier: Ambeed
|
Danger
|
A325093-100MG
A325093-1MG
A325093-50MG
76802-954EA
12.74
USD
76802-954
76802-956
76802-958
3-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)quinazoline-2,4(1H,3H)-dione (2R,3R)-2,3-dihydroxysuccinate 97%
3-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)quinazoline-2,4(1H,3H)-dione (2R,3R)-2,3-dihydroxysuccinate
Precaución: for Research Use Only
Formula:
C₂₆H₂₈FN₃O₉ MW: 545.51 g/mol |
MDL Number:
MFCD01530062 CAS Number: 83846-83-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...