3-Acetylcoumarin 98%

Supplier: Ambeed
Warning

A325032-1G A325032-5G
76802-950EA 12.74 USD
76802-950 76802-952
3-Acetylcoumarin 98%
3-Acetylcoumarin

Precaución: for Research Use Only

Formula: C₁₁H₈O₃
MDL Number: MFCD00006853
CAS Number: 3949-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR