Ethyl tetrahydro-2-furoate 98% (contains 3-5% EA)

Supplier: Ambeed
Warning

Synonyms: Ethyl tetrahydrofuran-2-carboxylate

A324539-1G A324539-250MG A324539-5G
76802-940EA 44.56 USD
76802-940 76802-942 76802-944
Ethyl tetrahydro-2-furoate 98% (contains 3-5% EA)
Ethyl tetrahydro-2-furoate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.168 g/mol
MDL Number: MFCD08063871
CAS Number: 16874-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR