H-DL-Asp(OMe)-OMe·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Asp(OMe)-OMe·HCl, H-DL-Asp(OMe)-OMe.HCl

A323485-10G A323485-500G A323485-5G A323485-100G A323485-25G
76802-934EA 12 USD
76802-934 76802-936 76802-938 77668-436 77668-438
H-DL-Asp(OMe)-OMe·HCl 97%
H-DL-Asp(OMe)-OMe·HCl

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₄
MW: 197.615 g/mol
MDL Number: MFCD00054368
CAS Number: 14358-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR