(2S,3S)-Methyl-2-amino-3-hydroxybutanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-Methyl 2-amino-3-hydroxybutanoate hydrochloride, H-allo-Thr-OMe·HCl

A322588-10G A322588-1G A322588-250MG A322588-25G A322588-5G
76802-920EA 885.86 USD
76802-920 76802-922 76802-924 76802-926 76802-928
(2S,3S)-Methyl-2-amino-3-hydroxybutanoate hydrochloride 97%
(2S,3S)-Methyl-2-amino-3-hydroxybutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₃
MW: 169.605 g/mol
MDL Number: MFCD00237766
CAS Number: 79617-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR