Potassium isobutyrate 98%

Supplier: Ambeed
Warning

Synonyms: Isobutyric Acid Potassium Salt

A319492-100G A319492-1G A319492-25G A319492-5G
76802-882EA 311 USD
76802-882 76802-884 76802-886 76802-888
Potassium isobutyrate 98%
Potassium isobutyrate

Precaución: for Research Use Only

Formula: C₄H₈O₂
MW: 126.20 g/mol
MDL Number: MFCD00058992
CAS Number: 19455-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR