3,4-Bis(benzyloxy)benzoic acid 97%

Supplier: Ambeed
Warning

A318895-10G A318895-1G A318895-25G A318895-5G
76802-848EA 115 USD
76802-848 76802-850 76802-852 76802-854
3,4-Bis(benzyloxy)benzoic acid 97%
3,4-Bis(benzyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₄
MDL Number: MFCD01861843
CAS Number: 1570-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR