5,6,7,8,9,10-Hexahydrocyclohept[b]indole 95%

Supplier: Ambeed
Warning

A317122-1G A317122-250MG A317122-5G
76802-840EA 24 USD
76802-840 76802-842 76802-844
5,6,7,8,9,10-Hexahydrocyclohept[b]indole 95%
5,6,7,8,9,10-Hexahydrocyclohept[b]indole

Precaución: for Research Use Only

Formula: C₁₃H₁₅N
MDL Number: MFCD00101340
CAS Number: 2047-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR