(±)-2-Methyl-2,4-pentanediol 98%

Supplier: Ambeed
Warning

Synonyms: Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol

A316830-100G A316830-500G A316830-1000G
76802-822EA 5.31 USD
76802-822 76802-824 77668-394
(±)-2-Methyl-2,4-pentanediol 98%
(±)-2-Methyl-2,4-pentanediol

Precaución: for Research Use Only

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
MDL Number: MFCD00004547
CAS Number: 107-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR