2-Naphthylacetic acid 97%

Supplier: Ambeed
Warning

A316668-100G A316668-10G A316668-25G A316668-500G A316668-5G
76802-812EA 64 USD
76802-812 76802-814 76802-816 76802-818 76802-820
2-Naphthylacetic acid 97%
2-Naphthylacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MDL Number: MFCD00004126
CAS Number: 581-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR