Ethylene glycol brassylate 98%

Supplier: Ambeed
Warning

A316068-100G A316068-500G
76802-806EA 19 USD
76802-806 76802-808 76802-810
Ethylene glycol brassylate 98%
Ethylene glycol brassylate

Precaución: for Research Use Only

Formula: C₁₅H₂₆O₄
MDL Number: MFCD00046977
CAS Number: 105-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR