Methoxyacetic acid hydrazide 95%

Supplier: Ambeed
Danger

Synonyms: Methoxyacetic hydrazide, 2-Methoxyacetohydrazide

A315436-10G A315436-25G
76802-792EA 13.8 USD
76802-792 76802-794
Methoxyacetic acid hydrazide 95%
Methoxyacetic acid hydrazide

Precaución: for Research Use Only

Formula: C₃H₈N₂O₂
MW: 104.11 g/mol
MDL Number: MFCD02197887
CAS Number: 20605-41-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR