Isovaleric acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid

A314672-100G A314672-10G A314672-500G A314672-25G
76802-786EA 24 USD
76802-786 76802-788 76802-790 77306-116
Isovaleric acid 98%
Isovaleric acid

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MDL Number: MFCD00002726
CAS Number: 503-74-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR