1,3,5-Triazine-2,4,6-triamine Poly phosphate (N%42-44%,P%12-14%)

Supplier: Ambeed
Warning

A310801-1000G A310801-100G A310801-500G
76802-752EA 46 USD
76802-752 76802-754 76802-756
1,3,5-Triazine-2,4,6-triamine Poly phosphate (N%42-44%,P%12-14%)
1,3,5-Triazine-2,4,6-triamine Poly phosphate

Precaución: for Research Use Only

Formula: C₃H₉N₆O₄P
MDL Number: MFCD00060248
CAS Number: 20208-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR