Fmoc-p-carboxy-phe(OtBu)-OH 98%

Supplier: Ambeed
Warning

A310244-100MG A310244-1G A310244-250MG A310244-5G A310244-10G A310244-25G
76802-740EA 14 USD
76802-740 76802-742 76802-744 76802-746 77550-864 77568-626
Fmoc-p-carboxy-phe(OtBu)-OH 98%
Fmoc-p-carboxy-phe(OtBu)-OH

Precaución: For research use only.

Formula: C₂₉H₂₉NO₆
MDL Number: MFCD01317016
CAS Number: 183070-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR