Ethyl-3-iodobenzoate 98%
Supplier: Ambeed
|
Warning
|
A309829-100G
A309829-10G
A309829-25G
A309829-500G
A309829-5G
76802-724EA
55
USD
76802-724
76802-726
76802-728
76802-730
76802-732
Ethyl-3-iodobenzoate 98%
Ethyl-3-iodobenzoate
Precaución: for Research Use Only
Formula:
C₉H₉IO₂ |
MDL Number:
MFCD00044769 CAS Number: 58313-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...