1-Hydroxy-2-naphthaldehyde 98%

Supplier: Ambeed
Warning

A308182-1G A308182-250MG
76802-708EA 63.66 USD
76802-708 76802-710
1-Hydroxy-2-naphthaldehyde 98%
1-Hydroxy-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈O₂
MW: 172.1800 g/mol
MDL Number: MFCD00092325
CAS Number: 574-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR