Propyl heptanoate 98%

Supplier: Ambeed
Warning

A307848-25G A307848-5G
76802-698EA 40 USD
76802-698 76802-700
Propyl heptanoate 98%
Propyl heptanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MW: 172.26 g/mol
MDL Number: MFCD00048897
CAS Number: 7778-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR