3-Amino-4-(4-morpholino)benzotrifluoride 97%

Supplier: Ambeed
Danger

A307730-1G A307730-250MG
76802-694EA 41 USD
76802-694 76802-696
3-Amino-4-(4-morpholino)benzotrifluoride 97%
3-Amino-4-(4-morpholino)benzotrifluoride

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃N₂O
MDL Number: MFCD00042160
CAS Number: 784-57-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR