(S)-4-Methyl-2-(methylamino)pentanoic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-D-Leu-OMe·HCl
A307465-1G
A307465-5G
A307465-25G
76802-684EA
22
USD
76802-684
76802-686
77879-613
(S)-4-Methyl-2-(methylamino)pentanoic acid hydrochloride 98%
(S)-4-Methyl-2-(methylamino)pentanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₆ClNO₂ MW: 181.66 g/mol |
MDL Number:
MFCD00754493 CAS Number: 66866-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...