Hexafluoropropene trimer 97%

Supplier: Ambeed
Danger

A306584-25G A306584-5G
76802-660EA 42 USD
76802-660 76802-662
Hexafluoropropene trimer 97%
Hexafluoropropene trimer

Precaución: for Research Use Only

Formula: C₉F₁₈
MDL Number: MFCD00043821
CAS Number: 6792-31-0
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR