1-Adamantanecarboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Adamantane-1-carboxamide

A306518-10G A306518-250MG A306518-5G
76802-654EA 151 USD
76802-654 76802-656 76802-658
1-Adamantanecarboxamide 97%
1-Adamantanecarboxamide

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MDL Number: MFCD00077200
CAS Number: 5511-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR