Methyl pyrrolidine-2-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Pro-OMe.HCl, H-DL-Pro-OMe·HCl

A306495-100G A306495-10G A306495-1G A306495-25G A306495-5G
76802-636EA 133.68 USD
76802-636 76802-638 76802-640 76802-642 76802-644
Methyl pyrrolidine-2-carboxylate hydrochloride 98%
Methyl pyrrolidine-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.617 g/mol
MDL Number: MFCD00066138
CAS Number: 79397-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR