Ethyl-2-amino-3-hydroxypropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-Ser-OEt·HCl, DL-serine ethyl ester hydrochloride

A306097-100G A306097-10G A306097-25G A306097-5G
76802-622EA 70.02 USD
76802-622 76802-624 76802-626 76802-628
Ethyl-2-amino-3-hydroxypropanoate hydrochloride 97%
Ethyl-2-amino-3-hydroxypropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₃·HCl
MW: 169.61 g/mol
MDL Number: MFCD00067544
CAS Number: 3940-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR