Isoamyl salicylate 97% (mixture of isomers)

Supplier: Ambeed
Danger

A305951-100G A305951-10G A305951-500G
76802-618EA 22 USD
76802-618 76802-620 77879-606
Isoamyl salicylate 97% (mixture of isomers)
Isoamyl salicylate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MDL Number: MFCD00020037
CAS Number: 87-20-7
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR