Methyl (R)-(+)-3-bromoisobutyrate 95%

Supplier: Ambeed
Warning

Synonyms: (R)-Methyl 3-bromo-2-methylpropanoate, Methyl (R)-(+)-3-Bromo-2-methylpropionate, (R)-(+)-3-Bromoisobutyric Acid Methyl Ester, (R)-(+)-3-Bromo-2-Methylpropionic Acid Methyl Ester

A305932-1G A305932-5G A305932-250MG
76802-614EA 49 USD
76802-614 76802-616 77668-354
Methyl (R)-(+)-3-bromoisobutyrate 95%
Methyl (R)-(+)-3-bromoisobutyrate

Precaución: for Research Use Only

Formula: C₅H₉BrO₂
MDL Number: MFCD00010639
CAS Number: 110556-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR