(S)-Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A304743-1G A304743-250MG A304743-5G
76802-598EA 50.93 USD
76802-598 76802-600 76802-602
(S)-Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%
(S)-Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₂
MW: 227.69 g/mol
MDL Number: MFCD05861484
CAS Number: 78183-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR