2-(Chloromethyl)-4-(3-methoxypropoxy)-3-methylpyridine hydrochloride 97%
Supplier: Ambeed
|
Danger
|
A303905-100G
A303905-10G
A303905-1G
A303905-25G
A303905-5G
76802-580EA
52
USD
76802-580
76802-582
76802-584
76802-586
76802-588
2-(Chloromethyl)-4-(3-methoxypropoxy)-3-methylpyridine hydrochloride 97%
Precaución: for Research Use Only
Formula:
C₁₁H₁₇Cl₂NO₂ |
MDL Number:
MFCD06658231 CAS Number: 153259-31-5 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...