Methyl-2,4-dihydroxy-3,6-dimethylbenzoate 98%

Supplier: Ambeed
Warning

A303174-100G A303174-1MG A303174-25G A303174-500G A303174-5G A303174-1000G A303174-100MG
76802-562EA 51 USD
76802-562 76802-564 76802-566 76802-568 76802-570 77879-596 77879-597
Methyl-2,4-dihydroxy-3,6-dimethylbenzoate 98%
Methyl-2,4-dihydroxy-3,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00157202
CAS Number: 4707-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR