2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone 90%

Supplier: Ambeed
Warning

A302471-100G A302471-25G A302471-5G
76802-554EA 342.68 USD
76802-554 76802-556 76802-558
2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone 90%
2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrFO
MW: 257.10 g/mol
MDL Number: MFCD11977644
CAS Number: 204205-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR