3-(2-Hydroxyethyl)-5-methoxyindole 97%

Supplier: Ambeed
Warning

A301743-100MG A301743-1G A301743-250MG
76802-536EA 76 USD
76802-536 76802-538 76802-540
3-(2-Hydroxyethyl)-5-methoxyindole 97%
3-(2-Hydroxyethyl)-5-methoxyindole

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD00047196
CAS Number: 712-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR