N,N-Dimethyl-1H-imidazole-1-sulfonamide 98%

Supplier: Ambeed
Warning

A300878-100G A300878-10G A300878-1G A300878-250MG A300878-25G A300878-5G
76802-524EA 524.09 USD
76802-524 76802-526 76802-528 76802-530 76802-532 76802-534
N,N-Dimethyl-1H-imidazole-1-sulfonamide 98%
N,N-Dimethyl-1H-imidazole-1-sulfonamide

Precaución: for Research Use Only

Formula: C₅H₉N₃O₂S
MW: 175.21 g/mol
MDL Number: MFCD00955713
CAS Number: 78162-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR