H-N-Me-Ala-OMe·HCl 98%

Supplier: Ambeed
Warning

A297079-1G A297079-250MG A297079-25G A297079-5G
76802-478EA 33 USD
76802-478 76802-480 76802-482 76802-484
H-N-Me-Ala-OMe·HCl 98%
H-N-Me-Ala-OMe·HCl

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·HCl
MDL Number: MFCD04973252
CAS Number: 20045-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR