4-Azaphthalide 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)nicotinic acid gamma-lactone, Furo[3,4-b]pyridin-5(7H)-one

A294815-1G A294815-5G
76802-430EA 98.67 USD
76802-430 76802-432
4-Azaphthalide 95%
4-Azaphthalide

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MW: 135.12 g/mol
MDL Number: MFCD01860224
CAS Number: 5657-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR