5-Methoxyindol-3-ylacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-3-indoleacetic acid, 2-(5-Methoxy-1H-indol-3-yl)acetic acid, 5-Methoxyindole-3-acetic acid

A293389-10G A293389-1G A293389-250MG A293389-25G A293389-5G
76802-412EA 195.21 USD
76802-412 76802-414 76802-416 76802-418 77601-306
5-Methoxyindol-3-ylacetic acid 95%
5-Methoxyindol-3-ylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
MDL Number: MFCD00005638
CAS Number: 3471-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR