Calcium-L-threonate 95%

Supplier: Ambeed
Warning

Synonyms: Calcium L-Threonate, L-Threonic acid Calcium salt, Calcium (2R,3S)-2,3,4-trihydroxybutanoate

A293097-5G A293097-25G A293097-500G A293097-100G A293097-1000G
76802-406EA 5 USD
76802-406 77601-302 77601-304 77668-300 77879-574
Calcium-L-threonate 95%
Calcium-L-threonate

Precaución: for Research Use Only

Formula: C₄H₈O₅
MW: 310.27 g/mol
MDL Number: MFCD00150824
CAS Number: 70753-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR