1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: 1-[2-(2,4-Dichlorophenyl)-2-hydroxyethyl]imidazole, 1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanol

A290204-1000G A290204-100G A290204-10G A290204-25G A290204-500G
76802-368EA 168.69 USD
76802-368 76802-370 76802-372 76802-374 76802-376
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol 97%
1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂N₂O
MW: 257.11 g/mol
MDL Number: MFCD00044708
CAS Number: 24155-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR