EDTA (ethylenediamine tetraacetic acid) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
A287527-100G
A287527-25G
A287527-500G
A287527-1000G
76802-326EA
8.49
USD
76802-326
76802-328
76802-330
77568-584
EDTA (ethylenediamine tetraacetic acid) 98%
EDTA (ethylenediamine tetraacetic acid)
Precaución: For research use only.
Formula:
C₁₀H₁₆N₂O₈ MW: 292.25 g/mol |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...