Heptaethylene glycol monododecyl ether 98%

Supplier: Ambeed
Danger

Synonyms: 3,6,9,12,15,18,21-Heptaoxatritriacontan-1-ol

A287490-100MG A287490-1G A287490-250MG A287490-5G
76802-318EA 48.81 USD
76802-318 76802-320 76802-322 76802-324
Heptaethylene glycol monododecyl ether 98%
Heptaethylene glycol monododecyl ether

Precaución: for Research Use Only

Formula: C₂₆H₅₄O₈
MW: 494.71 g/mol
MDL Number: MFCD00043065
CAS Number: 3055-97-8
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR