Fmoc-Asp(ODmab)-OH 98+%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asp(ODMAB)-OH

A287202-100MG A287202-1G A287202-250MG A287202-5G
76802-310EA 17 USD
76802-310 76802-312 76802-314 76802-316
Fmoc-Asp(ODmab)-OH 98+%
Fmoc-Asp(ODmab)-OH

Precaución: for Research Use Only

Formula: C₃₉H₄₂N₂O₈
MW: 666.771 g/mol
MDL Number: MFCD00797875
CAS Number: 269066-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR