Elaidic acid 97%

Supplier: Ambeed
Warning

A286651-1G A286651-25G A286651-5G
76802-308EA 43 USD
76802-308 77601-264 77601-266
Elaidic acid 97%
Elaidic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₂
MDL Number: MFCD00063954
CAS Number: 112-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR