Tetradecanal 97%

Supplier: Ambeed
Warning

Synonyms: Tetradecanaldehyde, Tetradecanal, Myristaldehyde, Tetradecyl aldehyde, Myristyl aldehyde

A285058-25G A285058-5G A285058-1G A285058-100MG A285058-250MG
76802-294EA 1101 USD
76802-294 76802-296 77550-752 77601-246 77601-248
Tetradecanal 97%
Tetradecanal

Precaución: for Research Use Only

Formula: C₁₄H₂₈O
MW: 212.38 g/mol
MDL Number: MFCD00007019
CAS Number: 124-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR