1-Boc-piperidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

A284839-1000G A284839-100G A284839-10G A284839-25G A284839-500G A284839-5G
76802-282EA 119 USD
76802-282 76802-284 76802-286 76802-288 76802-290 76802-292
1-Boc-piperidine-4-carboxylic acid 95%
1-Boc-piperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD00076999
CAS Number: 84358-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR